siRNA Details
| Gene Symbol: Fxyd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172317.2 | NP_758528.1 | 5 | 160 | ||
| XM_006228818.2 | XP_006228880.1 | 5 | 410 | |||
| XM_006228819.2 | XP_006228881.1 | 5 | 264 | |||
| XM_006228820.2 | XP_006228882.1 | 5 | 400 | |||
| GenBank mRNA | AW142066.1 | - | 4 | 165 | ||
| BG373727.1 | - | 4 | 307 | |||
| CA505583.1 | - | 4 | 306 | |||
| DY471861.1 | - | 5 | 532 | |||

Fxyd3
Genomic Map