siRNA Details
| Gene Symbol: Polr2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053948.4 | NP_446400.1 | 4 | 382 | ||
| XM_006230973.1 | XP_006231035.1 | 6 | 667 | |||
| GenBank mRNA | BC060550.1 | - | 4 | 338 | 6890594 | |
| BC060595.1 | - | 6 | 676 | 6888231 | ||
| FQ223906.1 | - | 6 | 672 | |||
| FQ224086.1 | - | 4 | 415 | |||
| FQ230390.1 | - | 4 | 345 | |||
| FQ232354.1 | - | 4 | 386 | |||
| Z71925.1 | - | 4 | 383 | |||

Polr2g
Genomic Map