siRNA Details
| Gene Symbol: Polr2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053948.4 | NP_446400.1 | 7 | 565 | ||
| XM_006230973.1 | XP_006231035.1 | 9 | 850 | |||
| GenBank mRNA | BC060550.1 | - | 7 | 521 | 6890594 | |
| BC060595.1 | - | 9 | 859 | 6888231 | ||
| FQ223906.1 | - | 9 | 855 | |||
| FQ230390.1 | - | 7 | 528 | |||
| FQ232354.1 | - | 7 | 569 | |||
| Z71925.1 | - | 7 | 566 | |||

Polr2g
Genomic Map