siRNA Details
| Gene Symbol: Ptf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053964.1 | NP_446416.1 | 1 | 296 | ||
| GenBank mRNA | X98170.1 | - | 1 | 288 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptf1a| Gene Symbol: Ptf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053964.1 | NP_446416.1 | 1 | 296 | ||
| GenBank mRNA | X98170.1 | - | 1 | 288 | ||