siRNA Details
| Gene Symbol: Rab28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053978.1 | NP_446430.1 | 1, 2 | 88 | ||
| XM_008770195.1 | XP_008768417.1 | 1, 2 | 457 | |||
| XM_008770196.1 | XP_008768418.1 | 1, 2 | 457 | |||
| XM_008770198.1 | XP_008768420.1 | 1, 2 | 457 | |||
| GenBank mRNA | BC072512.1 | - | 1, 2 | 212 | 7104824 | |
| X78606.1 | - | 1, 2 | 88 | |||

Rab28
Genomic Map