siRNA Details
| Gene Symbol: Pou2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228371.2 | XP_006228433.2 | 7, 8 | 612 | ||
| GenBank mRNA | L25863.1 | - | 7, 8 | 497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pou2f2| Gene Symbol: Pou2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228371.2 | XP_006228433.2 | 7, 8 | 612 | ||
| GenBank mRNA | L25863.1 | - | 7, 8 | 497 | ||