siRNA Details
Gene Symbol: Bhlhe41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729454.5 | XP_002729500.2 | 4 | 671 | ||
GenBank mRNA | AF009329.1 | - | 4 | 569 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bhlhe41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729454.5 | XP_002729500.2 | 4 | 671 | ||
GenBank mRNA | AF009329.1 | - | 4 | 569 |