siRNA Details
| Gene Symbol: Rab8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053998.2 | NP_446450.2 | 1 | 103 | ||
| GenBank mRNA | BC071176.1 | - | 1 | 96 | ||
| BC105863.1 | - | 1 | 103 | 7457017 | ||
| JN019799.1 | - | 1 | 98 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab8a| Gene Symbol: Rab8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053998.2 | NP_446450.2 | 1 | 103 | ||
| GenBank mRNA | BC071176.1 | - | 1 | 96 | ||
| BC105863.1 | - | 1 | 103 | 7457017 | ||
| JN019799.1 | - | 1 | 98 | |||