siRNA Details
| Gene Symbol: Sh3bp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054011.1 | NP_446463.2 | 5 | 556 | ||
| XM_008770976.1 | XP_008769198.1 | 3 | 2418 | |||
| GenBank mRNA | AB027562.1 | - | 5 | 556 | ||
| FQ210473.1 | - | 6 | 776 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3bp5| Gene Symbol: Sh3bp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054011.1 | NP_446463.2 | 5 | 556 | ||
| XM_008770976.1 | XP_008769198.1 | 3 | 2418 | |||
| GenBank mRNA | AB027562.1 | - | 5 | 556 | ||
| FQ210473.1 | - | 6 | 776 | |||