siRNA Details
| Gene Symbol: Slc15a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079838.1 | NP_001073307.1 | 2 | 264 | ||
| NM_057121.1 | NP_476462.1 | 22 | 1899 | |||
| XM_006252470.2 | XP_006252532.1 | 21 | 2140 | |||
| GenBank mRNA | AY860424.1 | - | 2 | 264 | ||
| D50306.1 | - | 22 | 1897 | |||
| D50664.1 | - | 22 | 1899 | |||

Slc15a1
Genomic Map