siRNA Details
| Gene Symbol: Khdrbs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130405.1 | NP_569089.1 | 1 | 389 | ||
| GenBank mRNA | AF305619.1 | - | 1 | 389 | ||
| AF393783.1 | - | 1 | 306 | |||
| BC061987.1 | - | 1 | 413 | 5599491 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Khdrbs1| Gene Symbol: Khdrbs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130405.1 | NP_569089.1 | 1 | 389 | ||
| GenBank mRNA | AF305619.1 | - | 1 | 389 | ||
| AF393783.1 | - | 1 | 306 | |||
| BC061987.1 | - | 1 | 413 | 5599491 | ||