siRNA Details
| Gene Symbol: Tff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057129.1 | NP_476470.1 | 2, 3 | 262 | ||
| GenBank mRNA | D83231.1 | - | 2, 3 | 262 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tff1| Gene Symbol: Tff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057129.1 | NP_476470.1 | 2, 3 | 262 | ||
| GenBank mRNA | D83231.1 | - | 2, 3 | 262 | ||