siRNA Details
| Gene Symbol: Rhoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057132.3 | NP_476473.1 | 2 | 195 | ||
| XM_006243699.1 | XP_006243761.1 | 4 | 463 | |||
| XM_006243700.1 | XP_006243762.1 | 4 | 438 | |||
| XM_006243701.1 | XP_006243763.1 | 4 | 558 | |||
| GenBank mRNA | AY026068.1 | - | 2 | 195 | ||
| AY026069.1 | - | 2 | 195 | |||
| BC061732.1 | - | 4 | 372 | 5598621 | ||
| D84477.1 | - | 2 | 165 | |||

Rhoa
Genomic Map