siRNA Details
Gene Symbol: Csrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057144.1 | NP_476485.1 | 3 | 367 | ||
XM_006229237.2 | XP_006229299.1 | 4 | 471 | |||
GenBank mRNA | X81193.1 | - | 3 | 367 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057144.1 | NP_476485.1 | 3 | 367 | ||
XM_006229237.2 | XP_006229299.1 | 4 | 471 | |||
GenBank mRNA | X81193.1 | - | 3 | 367 |