siRNA Details
| Gene Symbol: C9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057146.1 | NP_476487.1 | 8 | 1212 | ||
| GenBank mRNA | BC091127.1 | - | 8 | 1208 | 7324872 | |
| U49071.1 | - | 8 | 1194 | |||
| U52948.1 | - | 8 | 1212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9| Gene Symbol: C9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057146.1 | NP_476487.1 | 8 | 1212 | ||
| GenBank mRNA | BC091127.1 | - | 8 | 1208 | 7324872 | |
| U49071.1 | - | 8 | 1194 | |||
| U52948.1 | - | 8 | 1212 | |||