siRNA Details
| Gene Symbol: LOC103694886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773858.1 | XP_008772080.1 | 648 | |||
| Gene Symbol: C7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760831.1 | XP_008759053.1 | 9 | 1159 | ||

LOC103694886
Genomic Map