siRNA Details
| Gene Symbol: Chd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237395.2 | XP_006237457.1 | 18 | 2920 | ||
| XM_006237396.2 | XP_006237458.1 | 18 | 2920 | |||
| XM_006237397.2 | XP_006237459.1 | 18 | 2919 | |||
| XM_006237398.2 | XP_006237460.1 | 18 | 2919 | |||
| XM_006237399.2 | XP_006237461.1 | 18 | 2898 | |||
| XM_006237400.2 | XP_006237462.1 | 18 | 2880 | |||
| XM_006237401.2 | XP_006237463.1 | 18 | 2919 | |||
| XM_232354.8 | XP_232354.5 | 18 | 2898 | |||

Chd4
Genomic Map