siRNA Details
| Gene Symbol: Decr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057197.2 | NP_476545.2 | 3 | 349 | ||
| GenBank mRNA | BC059120.1 | - | 3 | 337 | 6920489 | |
| CO385941.1 | - | 3 | 334 | |||
| D00569.1 | - | 3 | 322 | |||
| FQ209689.1 | - | 3 | 338 | |||
| FQ210721.1 | - | 3 | 331 | |||
| FQ212507.1 | - | 3 | 327 | |||
| FQ218387.1 | - | 3 | 327 | |||
| FQ219629.1 | - | 3 | 330 | |||
| FQ219666.1 | - | 3 | 327 | |||
| FQ220227.1 | - | 3 | 330 | |||

Decr1
Genomic Map