siRNA Details
| Gene Symbol: Decr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057197.2 | NP_476545.2 | 10 | 1022 | ||
| GenBank mRNA | BC059120.1 | - | 10 | 1010 | 6920489 | |
| D00569.1 | - | 10 | 995 | |||
| FQ209689.1 | - | 10 | 1011 | |||
| FQ210607.1 | - | 10 | 1003 | |||
| FQ210721.1 | - | 10 | 1004 | |||
| FQ210830.1 | - | 6 | 599 | |||
| FQ212507.1 | - | 10 | 1000 | |||
| FQ218387.1 | - | 10 | 1000 | |||
| FQ219666.1 | - | 10 | 1000 | |||
| FQ220227.1 | - | 10 | 1003 | |||

Decr1
Genomic Map