siRNA Details
| Gene Symbol: Kif1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057200.2 | NP_476548.1 | 24 | 2626 | ||
| XM_008764204.1 | XP_008762426.1 | 24 | 2632 | |||
| XM_008764205.1 | XP_008762427.1 | 22 | 2336 | |||
| GenBank mRNA | AB070355.1 | - | 24 | 2626 | ||
| AB120429.1 | - | 21 | 2134 | |||

Kif1b
Genomic Map