siRNA Details
Gene Symbol: Tpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301286.1 | NP_001288215.1 | 2 | 230 | ||
NM_057208.2 | NP_476556.2 | 2 | 230 | |||
NM_173111.1 | NP_775134.1 | 2 | 272 | |||
XM_006232568.2 | XP_006232630.1 | 3 | 426 | |||
XM_006232569.2 | XP_006232631.1 | 3 | 426 | |||
XM_006232570.2 | XP_006232632.1 | 3 | 426 | |||
XM_006232571.2 | XP_006232633.1 | 3 | 426 | |||
XM_006232572.2 | XP_006232634.1 | 3 | 426 | |||
XM_006232573.2 | XP_006232635.1 | 3 | 426 | |||
XM_006232575.2 | XP_006232637.1 | 3 | 426 | |||
XM_006232576.2 | XP_006232638.1 | 3 | 426 | |||
XM_006232577.2 | XP_006232639.1 | 3 | 426 | |||
XM_006232580.2 | XP_006232642.1 | 2 | 330 | |||
XM_006232581.2 | XP_006232643.1 | 2 | 331 | |||
XM_006232583.2 | XP_006232645.1 | 2 | 334 | |||
XM_006232584.2 | XP_006232646.1 | 2 | 332 | |||
XM_008761124.1 | XP_008759346.1 | 2 | 330 | |||
XM_008761125.1 | XP_008759347.1 | 2 | 431 | |||
GenBank mRNA | FQ214881.1 | - | 3 | 404 | ||
FQ215752.1 | - | 3 | 406 | |||
FQ216429.1 | - | 3 | 405 | |||
FQ228755.1 | - | 2 | 280 | |||
L24775.1 | - | 2 | 230 | |||
L24776.1 | - | 230 | ||||
L24777.1 | - | 2 | 230 | |||
X72859.1 | - | 2 | 272 |
