siRNA Details
| Gene Symbol: Klf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057211.1 | NP_476559.1 | 1 | 769 | ||
| GenBank mRNA | D12769.1 | - | 1 | 769 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klf9| Gene Symbol: Klf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057211.1 | NP_476559.1 | 1 | 769 | ||
| GenBank mRNA | D12769.1 | - | 1 | 769 | ||