siRNA Details
| Gene Symbol: Lhx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139036.1 | NP_620605.1 | 2 | 608 | ||
| GenBank mRNA | L35572.1 | - | 2 | 608 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lhx5| Gene Symbol: Lhx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139036.1 | NP_620605.1 | 2 | 608 | ||
| GenBank mRNA | L35572.1 | - | 2 | 608 | ||