siRNA Details
| Gene Symbol: Rcan2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175578.3 | NP_783168.1 | 4 | 650 | ||
| XM_006244593.2 | XP_006244655.1 | 3 | 574 | |||
| GenBank mRNA | AF459022.1 | - | 3 | 372 | ||
| AF459023.1 | - | 4 | 492 | |||
| BC086974.1 | - | 4 | 650 | 7193921 | ||

Rcan2
Genomic Map