siRNA Details
| Gene Symbol: Baalc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144762.2 | NP_658907.1 | 3 | 643 | ||
| GenBank mRNA | AB073318.1 | - | 3 | 643 | ||
| AF371321.1 | - | 3 | 349 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Baalc| Gene Symbol: Baalc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144762.2 | NP_658907.1 | 3 | 643 | ||
| GenBank mRNA | AB073318.1 | - | 3 | 643 | ||
| AF371321.1 | - | 3 | 349 | |||