siRNA Details
| Gene Symbol: Cacng6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080694.1 | NP_542425.1 | 1 | 274 | ||
| XM_008758749.1 | XP_008756971.1 | 1 | 1354 | |||
| GenBank mRNA | AF361343.1 | - | 1 | 274 | ||
| AF361344.1 | - | 1 | 274 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacng6| Gene Symbol: Cacng6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080694.1 | NP_542425.1 | 1 | 274 | ||
| XM_008758749.1 | XP_008756971.1 | 1 | 1354 | |||
| GenBank mRNA | AF361343.1 | - | 1 | 274 | ||
| AF361344.1 | - | 1 | 274 | |||