siRNA Details
| Gene Symbol: Fabp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145878.1 | NP_665885.1 | 2 | 222 | ||
| GenBank mRNA | U13253.1 | - | 2 | 222 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fabp5| Gene Symbol: Fabp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145878.1 | NP_665885.1 | 2 | 222 | ||
| GenBank mRNA | U13253.1 | - | 2 | 222 | ||