siRNA Details
| Gene Symbol: Fabp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145878.1 | NP_665885.1 | 3 | 336 | ||
| XM_006232136.2 | XP_006232198.1 | 2 | 195 | |||
| GenBank mRNA | DQ255913.1 | - | 2 | 194 | ||
| FQ228381.1 | - | 3 | 348 | |||
| S69874.1 | - | 3 | 338 | |||
| S83247.1 | - | 3 | 353 | |||
| U13253.1 | - | 3 | 336 | |||

Fabp5
Genomic Map