siRNA Details
| Gene Symbol: Il2rg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080889.1 | NP_543165.1 | 3 | 446 | ||
| GenBank mRNA | AF410926.1 | - | 3 | 366 | ||
| AY325195.1 | - | 274 | ||||
| BC079343.1 | - | 3 | 446 | 7106866 | ||
| FQ232752.1 | - | 2 | 247 | |||

Il2rg
Genomic Map