siRNA Details
| Gene Symbol: Ikbkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080899.1 | NP_543175.1 | 17, 18 | 1986 | ||
| GenBank mRNA | AF388201.1 | - | 17, 18 | 1986 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ikbkap| Gene Symbol: Ikbkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080899.1 | NP_543175.1 | 17, 18 | 1986 | ||
| GenBank mRNA | AF388201.1 | - | 17, 18 | 1986 | ||