siRNA Details
| Gene Symbol: Arf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080904.2 | NP_543180.1 | 3 | 361 | ||
| XM_006257320.2 | XP_006257382.1 | 4 | 570 | |||
| GenBank mRNA | AY325223.1 | - | 4 | 328 | ||
| BC088865.1 | - | 3 | 361 | 7319260 | ||
| L12382.1 | - | 4 | 474 | |||

Arf3
Genomic Map