siRNA Details
Gene Symbol: Arf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080904.2 | NP_543180.1 | 3 | 421 | ||
XM_006257320.2 | XP_006257382.1 | 4 | 630 | |||
GenBank mRNA | AY325223.1 | - | 4 | 388 | ||
BC088865.1 | - | 3 | 421 | 7319260 | ||
L12382.1 | - | 4 | 534 |
