siRNA Details
| Gene Symbol: Paics | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080910.3 | NP_543186.1 | 5 | 518 | ||
| XM_006250852.2 | XP_006250914.1 | 5 | 566 | |||
| XM_006250853.2 | XP_006250915.1 | 5 | 537 | |||
| XM_006250854.2 | XP_006250916.1 | 5 | 567 | |||
| GenBank mRNA | BC072508.1 | - | 5 | 508 | 7103198 | |
| BC085711.1 | - | 5 | 518 | 7192781 | ||
| Gene Symbol: LOC100910308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751352.3 | XP_003751400.2 | 2 | 294 | ||

Paics
Genomic Map