siRNA Details
| Gene Symbol: Cfh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130409.2 | NP_569093.2 | 18 | 2966 | ||
| XM_008769531.1 | XP_008767753.1 | 17 | 2992 | |||
| GenBank mRNA | AF348145.1 | - | 3 | 456 | ||
| AJ320522.1 | - | 2940 | ||||
| CF110979.1 | - | 2 | 322 | |||

Cfh
Genomic Map