siRNA Details
Gene Symbol: Tmprss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130424.2 | NP_569108.2 | 6 | 643 | ||
XM_008768547.1 | XP_008766769.1 | 6 | 759 | |||
XM_008768548.1 | XP_008766770.1 | 6 | 690 | |||
XM_008768549.1 | XP_008766771.1 | 6 | 872 | |||
GenBank mRNA | AB073550.1 | - | 4 | 541 | ||
BC061712.1 | - | 6 | 643 | 5598010 |
