siRNA Details
| Gene Symbol: Eya2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130427.1 | NP_569111.1 | 2 | 114 | ||
| XM_008762456.1 | XP_008760678.1 | 3 | 350 | |||
| GenBank mRNA | AY366465.1 | - | 2 | 114 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eya2| Gene Symbol: Eya2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130427.1 | NP_569111.1 | 2 | 114 | ||
| XM_008762456.1 | XP_008760678.1 | 3 | 350 | |||
| GenBank mRNA | AY366465.1 | - | 2 | 114 | ||