siRNA Details
| Gene Symbol: Kcnk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130813.1 | NP_570826.1 | 1 | 322 | ||
| GenBank mRNA | AF467250.1 | - | 1 | 322 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnk15| Gene Symbol: Kcnk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130813.1 | NP_570826.1 | 1 | 322 | ||
| GenBank mRNA | AF467250.1 | - | 1 | 322 | ||