siRNA Details
Gene Symbol: Kcnk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130813.1 | NP_570826.1 | 2 | 626 | ||
GenBank mRNA | AF294353.1 | - | 2 | 305 | ||
AF467250.1 | - | 2 | 626 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcnk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130813.1 | NP_570826.1 | 2 | 626 | ||
GenBank mRNA | AF294353.1 | - | 2 | 305 | ||
AF467250.1 | - | 2 | 626 |