siRNA Details
| Gene Symbol: Psmd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130430.1 | NP_569114.1 | 2 | 650 | ||
| GenBank mRNA | AF067728.1 | - | 2 | 650 | ||
| FQ214178.1 | - | 194 | ||||
| FQ225583.1 | - | 183 | ||||
| FQ227319.1 | - | 183 | ||||
| FQ229304.1 | - | 216 | ||||
| Gene Symbol: LOC103694906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773903.1 | XP_008772125.1 | 1 | 387 | ||

Psmd9
Genomic Map