siRNA Details
| Gene Symbol: Psmd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130430.1 | NP_569114.1 | 3, 4 | 944 | ||
| GenBank mRNA | AF067728.1 | - | 3, 4 | 944 | ||
| FQ214178.1 | - | 488 | ||||
| FQ225583.1 | - | 477 | ||||
| FQ227319.1 | - | 477 | ||||
| Gene Symbol: LOC103694906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773903.1 | XP_008772125.1 | 2, 3 | 681 | ||

Psmd9
Genomic Map