siRNA Details
| Gene Symbol: Acaa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130433.1 | NP_569117.1 | 8 | 1096 | ||
| GenBank mRNA | FQ216726.1 | - | 8 | 959 | ||
| FQ228855.1 | - | 8 | 960 | |||
| X05341.1 | - | 8 | 1096 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acaa2| Gene Symbol: Acaa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130433.1 | NP_569117.1 | 8 | 1096 | ||
| GenBank mRNA | FQ216726.1 | - | 8 | 959 | ||
| FQ228855.1 | - | 8 | 960 | |||
| X05341.1 | - | 8 | 1096 | |||