siRNA Details
| Gene Symbol: Ifi27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203410.1 | NP_981955.1 | 6 | 598 | ||
| GenBank mRNA | BC060548.1 | - | 6 | 598 | 6919535 | |
| FQ219860.1 | - | 6 | 656 | |||
| FQ220098.1 | - | 6 | 667 | |||
| FQ226050.1 | - | 5 | 717 | |||
| FQ226998.1 | - | 6 | 631 | |||
| FQ230691.1 | - | 5 | 618 | |||
| FQ233730.1 | - | 6 | 632 | |||
| FQ233981.1 | - | 6 | 667 | |||

Ifi27
Genomic Map