siRNA Details
| Gene Symbol: Ifi27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203410.1 | NP_981955.1 | 2 | 101 | ||
| GenBank mRNA | AF154572.1 | - | 1 | 25 | ||
| BC060548.1 | - | 2 | 101 | 6919535 | ||
| FQ219860.1 | - | 2 | 159 | |||
| FQ220098.1 | - | 2 | 170 | |||
| FQ226050.1 | - | 1 | 220 | |||
| FQ226998.1 | - | 2 | 134 | |||
| FQ233730.1 | - | 2 | 135 | |||
| FQ233981.1 | - | 2 | 170 | |||

Ifi27
Genomic Map