siRNA Details
| Gene Symbol: Dmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759965.1 | XP_008758187.1 | 12 | 1128 | ||
| GenBank mRNA | AY548057.1 | - | 2023 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmbt1| Gene Symbol: Dmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759965.1 | XP_008758187.1 | 12 | 1128 | ||
| GenBank mRNA | AY548057.1 | - | 2023 | |||