siRNA Details
| Gene Symbol: Dmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022849.3 | NP_074040.2 | 2370 | |||
| XM_008759965.1 | XP_008758187.1 | 15 | 1425 | |||
| GenBank mRNA | AF540887.1 | - | 2370 | |||
| AY548057.1 | - | 2344 | ||||
| U32681.1 | - | 2063 | ||||

Dmbt1
Genomic Map