siRNA Details
| Gene Symbol: Dmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022849.3 | NP_074040.2 | 580 | |||
| XM_008759965.1 | XP_008758187.1 | 6 | 511 | |||
| GenBank mRNA | AF540887.1 | - | 580 | |||
| AY548057.1 | - | 464 | ||||
| U32681.1 | - | 273 | ||||
| Gene Symbol: LOC100911413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759957.1 | XP_008758179.1 | 8 | 854 | ||

Dmbt1
Genomic Map