siRNA Details
| Gene Symbol: Fgf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130752.1 | NP_570108.1 | 2 | 326 | ||
| GenBank mRNA | AB078901.1 | - | 2 | 326 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf21| Gene Symbol: Fgf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130752.1 | NP_570108.1 | 2 | 326 | ||
| GenBank mRNA | AB078901.1 | - | 2 | 326 | ||