siRNA Details
Gene Symbol: Acot8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130756.3 | NP_570112.3 | 3, 4 | 890 | ||
GenBank mRNA | AF452100.1 | - | 3, 4 | 540 | ||
BC078751.1 | - | 3, 4 | 884 | 7114090 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Acot8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130756.3 | NP_570112.3 | 3, 4 | 890 | ||
GenBank mRNA | AF452100.1 | - | 3, 4 | 540 | ||
BC078751.1 | - | 3, 4 | 884 | 7114090 |