siRNA Details
| Gene Symbol: Lhx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001059910.4 | XP_001059910.2 | 6 | 980 | ||
| XM_002729144.4 | XP_002729190.1 | 6 | 1594 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lhx3| Gene Symbol: Lhx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001059910.4 | XP_001059910.2 | 6 | 980 | ||
| XM_002729144.4 | XP_002729190.1 | 6 | 1594 | |||