siRNA Details
| Gene Symbol: Kcnq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133322.1 | NP_579856.1 | 9, 10 | 1274 | ||
| XM_006235721.2 | XP_006235783.1 | 9, 10 | 1358 | |||
| XM_006235727.2 | XP_006235789.1 | 9, 10 | 1357 | |||
| XM_006235728.2 | XP_006235790.1 | 9, 10 | 1357 | |||
| XM_008762457.1 | XP_008760679.1 | 9, 10 | 1358 | |||
| XM_008762459.1 | XP_008760681.1 | 9, 10 | 1358 | |||
| XM_008762461.1 | XP_008760683.1 | 9, 10 | 1358 | |||
| XM_008762462.1 | XP_008760684.1 | 9, 10 | 1357 | |||
| GenBank mRNA | AF087453.2 | - | 9, 10 | 1274 | ||

Kcnq2
Genomic Map